Learn More
Cole-Parmer™ Heavy Duty PS Pinch Clamp Closure for Tubing and Hose
Handle higher working pressures with these heavy-duty tubing closures. Cole-Parmer™ Heavy Duty Closures provide excellent durability, can be continually reused and resist most chemicals.
Brand:  Cole-Parmer™ 20001-00
Product Code. 16477770
  • 85.25 € / Each
Only null left
In Stock
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • High-strength polysulfone clamp
  • Quick installation with positive  swing-away locking screw
  • Smooth contours allow for easy cleaning
  • Simply align the hose within the clamp; mating surfaces on the clamp indicate complete closure 
  • Typical applications include sterile filling, media bag connections, pumping, liquid control and high-purity fluid transfers
  • FDA-compliant polysulfone material can be sterilized by autoclave or gamma radiation