Learn More
Thermo Scientific™ HEG Phosphoramidite, standard grade, serum vial bottle
Shop All Thermo Scientific ProductsDescription
HEG CEP (Spacer 18) is a hexaethyleneglycol-based phosphoramidite for use in oligonucleotide synthesis. This unique property creates a hydrophilic spacer, which can be useful for building flexibility within an oligo sequence. It has proven useful for probes requiring minimal steric hindrance (usually when conjugated to an antibody or enzyme), multiple binding sites, and polymerase activity at the 3' terminus to be eliminated. This product is supplied in a 20 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.
Features:
• Full chemical name: 18-O-(4,4'-Dimethoxytrityl)-Hexaethyleneglycol-1-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C42H61N2O10P
• Formula weight: 784.9 g/mol
• HPLC Purity: ≥95.0%; Bis impurity ≤0.3%; Penta impurity ≤1.6%; Hepta impurity ≤0.9%
• 31P NMR Purity: ≥98%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%
Specifications
Specifications
| For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
| Product Type | HEG CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | None (Modifier) |
| Shipping Condition | Dry Ice |
| Purity | 95% |
| Quantity | 1 g |
| Reagent Type | Phosphoramidite |
| Grade | Standard |
| Nucleic Acid Type | None (Modifier) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.