missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hegsted Salt Mixture

Product Code. 11455642 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455642 2 kg 2kg
11465642 10 kg 10kg
11475642 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455642 Supplier MP Biomedicals™ Supplier No. 0290284002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hegsted Salt Mixture is used for rodent diet.

  • Mineral Mix
  • For rodent diet
  • Powder
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
For Use With (Application) Rodent diet
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.