missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hegsted Salt Mixture

Product Code. 11475642 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475642 50 kg 50kg
11455642 2 kg 2kg
11465642 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475642 Supplier MP Biomedicals™ Supplier No. 0290284050
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hegsted Salt Mixture is used for rodent diet.

  • Mineral Mix
  • For rodent diet
  • Powder

Specifications

Product Type Animal Diet
For Use With (Application) Rodent diet
Quantity 50 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.