Learn More
Heidolph™ Hei-PLATE Silver 2 Package
Product Code. 17333440 Shop All Heidolph Instruments Products
Click to view available options
Plug Type:
CH-Style
Unit Size:
Each
Product Code. 17333440
Brand: Heidolph™ 5054008004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Package includes magnetic stirrer Hei-Connect, temperature sensor Pt1000, clamping system, and interface cable RS 232.
This package includes:
  • Magnetic Stirrer Hei-Connect
  • Temperature sensor Pt1000 (AISI 316Ti)
  • Pt1000 clamping system (includes support rod and attachment with cable inlet)
  • Interface cable RS 232
TRUSTED_SUSTAINABILITY
Specifications
Plug Type CH-Style