Learn More
Heidolph™ Hei-VAP Automatic Evaporation Package, UK-plug
Product Code. 17383110 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
Product Code. Plug Type Unit Size
17383110 UK-plug Each
1 options
Product Code. 17383110 Supplier Heidolph™ Supplier No. 5743730003
Estimated Shipment: 08-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Hei-VAP Automatic Evaporation Package features a motor lift, condenser, Woulff bottle, and tubing set.
  • Hei-VAP Expert Control with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Vario Tec
  • Condenser for Rotavac Vario
  • Tubing set
  • Woulff bottle
  • AUTOaccurate-sensor
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type UK-plug