Learn More
Heidolph™ Hei-VAP Core All-round Package, UK-plug
Product Code. 17363050 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
Product Code. Plug Type Unit Size
17363050 UK-plug Each
1 options
Product Code. 17363050 Supplier Heidolph™ Supplier No. 5712030003
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Hei-VAP Core All-round Package features a hand lift, Hei-VAC Control, vacuum pump, and tubing set.
  • Hei-VAP Core with hand lift and vertical glassware set G3
  • Hei-VAC Control
  • Vacuum pump Rotavac Valve Tec
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Type Hand Lift Rotary Evaporator
Plug Type UK-plug