missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Hei-VAP Powder Drying-Chill Package, UK-plug

Product Code. 17373110 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
17373110 UK-plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17373110 Supplier Heidolph™ Supplier No. 5743330003
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Hei-VAP Powder Drying-Chill Package features a motor lift, chiller, powder flask, and tubing set.

  • Hei-VAP Expert Control with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Valve Control
  • Vacuum valve
  • Chiller Hei-CHILL 600
  • RS 232 cable
  • Tubing set
  • Powder flask 2 L
  • Vapor tube with powder filter
  • Thermal fluid Kyro 30 (10 L)
TRUSTED_SUSTAINABILITY

Specifications

Type Motor Lift Rotary Evaporator
Plug Type UK-plug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.