Learn More
Heidolph™ Hei-VAP Reaction Extraction-Chill Package, UK-plug
Product Code. 17343110 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
Product Code. Plug Type Unit Size
17343110 UK-plug Each
1 options
Product Code. 17343110 Supplier Heidolph™ Supplier No. 5742360003
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Hei-VAP Reaction Extraction-Chill Package features a motor lift, chiller, and vapor temperature sensor.
  • Hei-VAP Expert Control with motor lift and vertical glassware set G6
  • Vacuum pump Rotavac Vario Tec
  • Vapor temperature sensor
  • Chiller Hei-CHILL 350
  • RS 232 cable
  • Thermal fluid Kyro 30 (10 L)
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type UK-plug