Learn More
Heidolph™ Hei-VAP University Academic-Chill Package, UK-plug
Product Code. 17393050 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
Product Code. Plug Type unitSize
17393050 UK-plug Each
1 options
Product Code. 17393050 Supplier Heidolph™ Supplier No. 5712510003
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
The Hei-VAP University Academic-Chill Package features a hand lift, vacuum pump, chiller, and tubing set.
  • Hei-VAP Core with hand lift and diagonal glassware set G1
  • Vacuum pump Rotavac Valve Tec
  • Manual vacuum controller
  • Chiller Hei-CHILL 250
  • Thermal fluid Kyro 30 (10 L)
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Type Hand Lift Rotary Evaporator
Plug Type UK-plug