Learn More
Heidolph™ Hei-VAP University Research-Chill Package, UK-plug
Product Code. 17313160 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
UK-plug
Unit Size:
Each
Product Code. Plug Type Unit Size
17313160 UK-plug Each
1 options
Product Code. 17313160 Supplier Heidolph™ Supplier No. 5762730503
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Hei-VAP University Research-Chill Package features a motor lift, chiller, and tubing set.
  • Hei-VAP Ultimate Control with motor lift and vertical glassware set G3 XL
  • Vacuum pump Rotavac Vario Control
  • Chiller Hei-CHILL 600
  • RS 232 cable
  • Tubing set
  • Thermal fluid Kyro 30 (10 L)
TRUSTED_SUSTAINABILITY
Specifications
Type Motor Lift Rotary Evaporator
Plug Type UK-plug