missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Helico Blot 2.1 (RUO)

Product Code. 30694939 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
18 Tests/Kit
36 Tests/Kit
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30694939 18 Tests/Kit 1 set
30694934 36 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30694939 Supplier MP Biomedicals Supplier No. 12422018
  • ONLINE EXCLUSIVE PRICE
    505.98 €
    481.89 € / 1 sets
    Save 24.09 €
    4% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Blot is a qualitative enzyme immunoassay for the in vitro detection of Helicobacter pylori antibodies in human serum or plasma.

  • Nitrocellulose strips are precoated with H. pylori antigens and with a recombinant antigen of H. pylori.
  • Individual strips are incubated with diluted serum or plasma specimens or controls.
  • Specific antibodies to the various antigens, if present, will bind to the H. pylori antigens on the strips respectively.
  • The strips are washed to remove unbound antibodies. Specifically bound antibodies are visualized using a series of reaction with the goat anti-human IgG conjugated with alkaline phosphatase and the substrate BCIP/NBT.
  • The test allows the user to differentiate the reactivity to each of the various H.pylori antigens.
  • The product is intended for use as a serological test for the detection of both current and past infection with H. pylori.
  • Research Use Only (RUO).
  • For professional use only.
  • Human serum and plasma.

Specifications

Product Type Western Blot Kit
Detection Method Colorimetric
For Use With (Application) Immunoblot (Western)
Membrane Compatibility Nitrocellulose
Quantity 18 Tests/Kit
Substrate Alkaline Phosphatase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.