Learn More
Eppendorf™ Helix Chambers
Product Code. 12836532 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Helix Fusion Chambers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12836532 Helix Fusion Chambers Each
1 options
Product Code. 12836532 Supplier Eppendorf™ Supplier No. 4308017007
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Extremely high transformation rates due to innovative electronics and buffers
  • Extremely high transformation rates due to innovative electronics and buffers
  • Integrated cuvette chamber
  • Easy to use
  • Compact design
  • Universal electroporator for eukaryotic, yeast and bacteria
TRUSTED_SUSTAINABILITY
Specifications
Type Helix Fusion Chamber Stand
For Use With (Equipment) Helix Fusion Chambers
Holds 10
Quantity 1 Each