Learn More
BRAND™ Hellendahl Extended Staining Trough
Product Code. 10232001 Shop All Brand Products
Change view
Click to view available options
Slide Count:
16
Unit Size:
Pack of 4
Product Code. Slide Count Unit Size
10232001 16 Pack of 4
1 options
Product Code. 10232001 Supplier BRAND™ Supplier No. 474400
  • ONLINE EXCLUSIVE PRICE
    104.00 €
    66.67 € / Pack of 4
    Save 37.33 €
    35% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
PMP, transparent, for 16 slides
  • For 16 slides
    • 76 x 26 mm
    • 8 pairs back-to-back
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material PMP
Type Hellendahl Extended Staining Trough
Slide Count 16