missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Hematocrit Tubes, NH4-heparinized

Product Code. 10595592
Change view
Click to view available options
Capacity (Metric):
18 μL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10595592 18 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10595592 Supplier Hirschmann™ Supplier No. 9100418

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Excellent for centrifuging blood in the laboratory. Hirschmann™ Hematocrit Tubes are manufactured from disposable capillary glass.

  • Disposable
  • With ringmark at 60mm
  • NH4-heparinized
  • Color-code green
  • CE acc. IVD 98/79 EC

Specifications

Product Type Hematocrit Tube NH4-hep
Capacity (Metric) 18 μL
Diameter (Metric) Inner 0.5/0.6 mm
For Use With (Application) For centrifuging blood samples, Ammonium heparinized
Length (Metric) 75 mm
Certifications/Compliance CE, IVD 98/79 EC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.