missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Sodium Heparinized Hematocrit Tubes

Product Code. 30090387
Change view
Click to view available options
Capacity (Metric):
60 μL
75 μL
100 μL
Unit Size:
Pack of 1000
Pack of 5000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30090387 75 μL Pack of 5000
10233391 100 μL Pack of 1000
30090390 100 μL Pack of 5000
10410772 60 μL Pack of 1000
30090388 60 μL Pack of 5000
10199641 75 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30090387 Supplier Hirschmann™ Supplier No. 9100275

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Sodium Heparinized Hematocrit Tubes are manufactured from disposable capillary glass.

  • Color-coded red
  • Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC

Specifications

Product Type Hematocrit Tube Na-hep
Capacity (Metric) 75 μL
Diameter (Metric) Inner 1.07/1.23 mm
For Use With (Application) For centrifuging blood samples, Sodium heparinized
Length (Metric) 75 ±0.5 mm
Certifications/Compliance DIN EN ISO 12772, CE, IVD 98/79 EC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.