missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Hematocrit Tubes

Product Code. 10146260
Change view
Click to view available options
Capacity (Metric):
60 μL
75 μL
Unit Size:
Pack of 1000
Pack of 5000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10146260 60 μL Pack of 1000
30090386 60 μL Pack of 5000
30090389 75 μL Pack of 5000
10604162 75 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10146260 Supplier Hirschmann™ Supplier No. 9100160

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Hematocrit Tubes are manufactured from disposable capillary glass.

  • Not heparinized
  • Color-coded blue
  • Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC

Specifications

Product Type Hematocrit Tube
Capacity (Metric) 60 μL
Diameter (Metric) Inner 0.9/1.0 mm
For Use With (Application) For centrifuging blood samples, not heparinized
Length (Metric) 75 ±0.5 mm
Certifications/Compliance DIN EN ISO 12772, CE, IVD 98/79 EC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.