Learn More
Tocris Bioscience™ Hematoxylin
Histologic stain; used to stain cell nuclei
Brand:  Tocris Bioscience™ 5222/25G
Encompass
Product Code. 15786655
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Hematoxylin is a dye used to stain cell nuclei. Hematoxylin is commonly combined with eosin for H&E histological staining and is a component of the Papanicolaou stain used in cytology. Hematoxylin can be used with other fixatives to stain muscle striations, fibrin, myelin and elastin. Can also be used for H&E staining of intact tissues.
Specifications
Specifications
Histology Stains
517-28-2
25 g
(6 AS,11 bR)-7,11 b-Dihydrobenz[b]indeno[1,2-d]pyran-3,4,6 A,9,10(6 H)-pentol
Room Temperature