missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Hematoxylin

Product Code. 18722448
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722448 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722448 Supplier Tocris Bioscience™ Supplier No. 5222/25G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Histologic stain; used to stain cell nuclei

Hematoxylin is a dye used to stain cell nuclei. Hematoxylin is commonly combined with eosin for H&E histological staining and is a component of the Papanicolaou stain used in cytology. Hematoxylin can be used with other fixatives to stain muscle striations, fibrin, myelin and elastin. Can also be used for H&E staining of intact tissues.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Histology Stains
Molecular Formula (6 AS,11 bR)-7,11 b-Dihydrobenz[b]indeno[1,2-d]pyran-3,4,6 A,9,10(6 H)-pentol
CAS 517-28-2
Content And Storage Room Temperature
Quantity 25 g
Product Type Hematoxylin
Molecular Weight (g/mol) M.W. (Observed): 302.28
Inhibitors Hematoxylin

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.