missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hematoxylin

Product Code. 16452499
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452499 Supplier MedChemExpress Supplier No. HYN0116
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hematoxylin (Natural Black 1), a naturally occurring flavonoid compound derived from the logwood tree, Haematoxylon campechianum.

Hematoxylin is a nuclear stain in histology and is also a potent Aβ42 fibrillogenesis inhibitor with an IC50 of 1.6 μM.

Specifications

Quantity 1 g
Storage Requirements Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Solubility DMSO : 50 mg/mL (165.41 mM; Need ultrasonic) ∣ H2O : 6.67 mg/mL (22.07 mM; Need ultrasonic)
Shipping Condition Room temperature
CAS 517-28-2
Molecular Weight (g/mol) 302.28 g/mol
Synonym Natural Black 1 ∣ Haematoxylin
Target NO Synthase ∣ MMP ∣ Endogenous Metabolite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.