missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hematoxylin, 85%, pure, certified

Product Code. 10548512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10548512 25 g Glass Bottle 25g
10239600 100 g Glass Bottle 100g
11966921 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10548512 Supplier Thermo Scientific Chemicals Supplier No. 411160250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 517-28-2 | C16H14O6 | 302.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black 1, C.I. 75290
PubChem CID 45029742
IUPAC Name (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

Melting Point 100.0°C to 120.0°C
Infrared Spectrum Authentic
Loss on Drying 10% (105°C) max.
Packaging Glass Bottle
Assay Percent Range 85%
UV_Vis 88% (on dry substance) min.
Color Beige to Brown
Solubility Information Solubility in water: slightly soluble; soluble in hot water;. Other solubilities: soluble in hot alcohol, glycerol, borax, alkali hy, droxides (aq. solution purple red; becomes blue vi, olett during storage at air).
Quantity 25 g
Physical Form Powder
Formula Weight 302.27
Percent Purity 85%
Grade Pure
Chemical Name or Material Hematoxylin
Show More Show Less
compliance-icons
Product Identifier
  • Hematoxylin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hematoxylin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.