missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin, 85%, pure, Thermo Scientific™

Product Code. 10772782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772782 100 g Glass bottle 100g
10166940 25 g Glass bottle 25g
10003973 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10772782 Supplier Thermo Scientific Acros Supplier No. 411181000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black 1, C.I. 75290
PubChem CID 45029742
IUPAC Name (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

Assay Percent Range 85%
Merck Index 15, 4672
Infrared Spectrum Authentic
Formula Weight 302.27
Loss on Drying 10 % max. (1 g, 105°C, 3 hrs)
Grade Pure
Packaging Glass bottle
UV_Vis 80 % min.
Color Beige to Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material Hematoxylin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.