missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin Trihydrate, Fisher BioReagents

Product Code. 10072594
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Amber Glass
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10072594 100 g Amber Glass 100g
10012694 5 g Amber Glass 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10072594 Supplier Fisher BioReagents Supplier No. BP2523100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Hematoxylin Trihydrate is used mainly as a stain in microscopy.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black I
PubChem CID 45029742
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

Melting Point 100°C
Merck Index 15, 4672
Identification Pass Test
IUPAC Name (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
Lambda Max 290 to 292nm
Formula Weight 356.4
Loss on Drying Report %
Packaging Amber Glass
Color Amber
Quantity 100 g
Physical Form Powder
Chemical Name or Material Hematoxylin Trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.