missing translation for 'onlineSavingsMsg'
Learn More

Hemogard™ Lavender Closure Blood Tube

Product Code. p-8000829
Click to view available options
Capacity (Metric):
3 mL
4 mL
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12316949

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Specifications

Product Type Blood Collection Tube
Additive K2 EDTA
Closure Type Hemogard
Capacity (Metric) 3 mL
Closure Color Lavender
Height (Metric) 75 mm
Diameter (Metric) Outer 13 mm
compliance-icons
Product Identifier
  • TUBE BLOOD EDTA / PLUS SPRAY COAT (K2) BL hemogard closure colour code lavender 13 x 75mm
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Dipotassium dihydrogen ethylenediaminetetraacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.