missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepaRG™ Cells, Cryopreserved

Product Code. 13496438 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13496438 1 vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13496438 Supplier Gibco™ Supplier No. HPRGC10
  • ONLINE EXCLUSIVE PRICE
    998.00 €
    898.65 € / Pack of 1
    Save 99.35 €
    9% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The HepaRG™ Cells, Cryopreserved cell line is an immortalized hepatic cell line that retains many characteristics of primary human hepatocytes.

The HepaRG™ cell line is an immortalized hepatic cell line that retains many characteristics of primary human hepatocytes. HepaRG™ cells are terminally differentiated and provided in a convenient cryopreserved format. For scientists who need reproducible metabolism data, HepaRG™ cells are an in vitro tool that provides reproducible results in a metabolically complete and scalable system.

  • Obtain biologically relevant results from a metabolically complete system
  • Assess the drug-drug interaction potential of your compound
  • Experience reproducible results from a single population of cells

Biologically Relevant

HepaRG™ cells exhibit many characteristics of primary human hepatocytes including morphology, expression of key metabolic enzymes, expression of nuclear receptors, and drug transporters. Unlike HepG2 and Fa2N-4 cells, HepaRG™ cells have high P450 activity and complete expression of all nuclear receptors.

Predict Metabolism-based Drug-drug Interaction

HepaRG™ cells respond to prototypical P450 inducers and inhibitors to the same extent as primary hepatocytes, allowing HepaRG™ cells to be used to evaluate potential drug-drug interactions.

Infinitely Reproducible

HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

HepaRG™ is a trademark of BioPredic International.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 vial, store in cryostorage dewar or -135°C freezer
Age Adult
Cell Line HepaRG
Cell Type Hepatocytes
Culture Type Hepatocyte Cell Culture
Form Cryopreserved
Gender Female
Species Human
Tissue Liver
Donor Source Single Donor
No. of Cells 10 x 106
Product Type Hepatocytes
Quantity 1 vial
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.