missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepaRG™ Maintenance/Metabolism Medium Supplement (5X)

Product Code. 11311870 Shop All Gibco Products
Change view
Click to view available options
Quantity:
76.7 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311870 76.7 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11311870 Supplier Gibco™ Supplier No. HPRG720
  • ONLINE EXCLUSIVE PRICE
    404.00 €
    371.65 € / 79mL
    Save 32.35 €
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HepaRG™ Maintenance/Metabolism Medium Supplement (5X) is a specialized media supplement for use in culturing HepaRG™ cells.

HepaRG™ Maintenance/Metabolism Medium Supplement (5x) is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Maintenance/Metabolism Medium Supplement (5x) is specifically used for multiple day growth of HepaRG™ cells.

  • Easy media preparation, just thaw, mix, and use
  • Reproducible, and biologically relevant results with HepaRG™ cells

Media Preparation

Working media is prepared by the addition of thawed HepaRG™ Maintenance/Metabolism Medium Supplement (5x) to 500 ml of William's Medium E (A1217601 or 12551032) and 5 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.

Simplified, Relevant and Reproducible Studies

HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

HepaRG™ is a trademark of BioPredic International.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Supplement should be stored at -20°C
Cell Type HepaRG Cells
Concentration 5X
Form Frozen
Product Type Supplement
Serum Level Standard Serum
Sterility Sterile
Without Additives No Glutamine
Format Bottle
Product Line Gibco
Purity or Quality Grade Research Grade, Cell Culture Grade
Quantity 76.7 mL
Shipping Condition Dry Ice
Source Human
Tested For Sterility Testing (Bacteria & Fungi)
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.