missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepaRG™ Maintenance/Metabolism Medium Supplement

Product Code. 11391840 Shop All Gibco Products
Change view
Click to view available options
Quantity:
16 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391840 16 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11391840 Supplier Gibco™ Supplier No. HPRG620
  • ONLINE EXCLUSIVE PRICE
    116.00 €
    112.65 € / 16mL
    Save 3.35 €
    2% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HepaRG™ Maintenance/Metabolism Medium Supplement

HepaRG™ Maintenance/Metabolism Medium Supplement is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Maintenance/Metabolism Medium Supplement is specifically used for multiple day growth of HepaRG™ cells.
  • Easy media preparation, just thaw, mix, and use
  • Reproducible, and biologically relevant results with HepaRG™ cells

    Media Preparation
    Working media is prepared by the addition of thawed HepaRG™ Maintenance/Metabolism Medium Supplement to 100 ml of William’s Medium E (A1217601 or 12551032) and 1 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.

    Simplified, Relevant and Reproducible Studies
    HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

    HepaRG™ is a trademark of BioPredic International.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Supplement should be stored at -20°C
    Cell Type HepaRG Cells
    Form Frozen
    Product Type Hepatocyte Supplement
    Serum Level Standard Serum
    Sterility Sterile
    Without Additives No Glutamine
    For Use With (Application) Drug Metabolism
    Product Line Gibco
    Purity or Quality Grade Research Grade, Cell Culture Grade
    Quantity 16 mL
    Shipping Condition Dry Ice
    Tested For Sterility Testing (Bacteria & Fungi)
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.