missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepaRG™ Thaw, Plate & General Purpose Medium Supplement (5X)

Product Code. 11371870 Shop All Gibco Products
Change view
Click to view available options
Quantity:
69 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371870 69 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11371870 Supplier Gibco™ Supplier No. HPRG770
  • ONLINE EXCLUSIVE PRICE
    404.00 €
    363.65 € / 67mL
    Save 40.35 €
    9% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

HepaRG™ Thaw, Plate & General Purpose Medium Supplement (5X) is a specialized media supplement for use in culturing HepaRG™ cells.

HepaRG™ Thaw, Plate, & General Purpose Medium Supplement (5x) is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Thaw, Plate, & General Purpose Medium Supplement (5x) is specifically used for the initial thawing and propagation of cryopreserved HepaRG™ cells.

  • Easy media preparation, just thaw, mix, and use
  • Reproducible, and biologically relevant results with HepaRG™ cells

Media Preparation

Working media is prepared by the addition of thawed HepaRG™ Thaw, Plate, & General Purpose Medium Supplement (5x) to 500 ml of William's Medium E (A1217601 or 12551032) and 5 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.

Simplified, Relevant and Reproducible Studies

HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

HepaRG™ is a trademark of BioPredic International.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Supplement should be stored at -20°C
Cell Type HepaRG Cells
Concentration 5X
Form Frozen
Product Type Hepatocyte Supplement
Serum Level Standard Serum
Sterility Sterile
Without Additives No Glutamine
Format Bottle
pH 6 to 8
Product Line Gibco
Purity or Quality Grade Research Grade, Cell Culture Grade
Quantity 69 mL
Shipping Condition Dry Ice
Tested For Sterility Testing (Bacteria & Fungi)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.