missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS Heparin Ammonium Salt

Product Code. 30928580 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30928580 250 mg 250mg
30928575 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30928580 Supplier MP BIOMEDICALS Supplier No. 0219411083
  • ONLINE EXCLUSIVE PRICE
    106.00 €
    99.75 € / 250mg
    Save 6.25 €
    5% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Heparin is a polymer classified as a mucopolysaccharide or glycosoaminoglycan, biosynthesized in mast cells of mammalian tissues.

  • Enhances anticoagulant activity by binding to antithrombin III, accelerating the inhibition of coagulation proteases such as factor Xa and thrombin.
  • Widely used in research and manufacturing applications to study blood coagulation and anticoagulation mechanisms.
  • Supplied for laboratory and industrial use only; not intended for direct human use.
  • Derived from mammalian tissues and carefully processed to preserve biological activity for experimental applications.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 250 mg
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.