missing translation for 'onlineSavingsMsg'
Learn More

Abcam Heparin Beads 6FF

Product Code. 32119833
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
100 mL
Unit Size:
100mL
1mL
25mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32119833 1 mL 1mL
32111688 5 mL 5mL
32112566 100 mL 100mL
32115482 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 32119833 Supplier Abcam Supplier No. ab3190951mL
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for purification of heparin-dependent biomolecules e.g. antithrombin III, coagulation factors and other plasma proteins.

Supplemental Information

  • Ligand: Heparin
  • Basic Beads: 6% Cross-linked agarose beads
  • Particle size: 45μm - 165μm
  • Storage Buffer: 80% PBS, 20% ethanol
  • pH Stability Range: 4-12
  • Maximum pressure: 0.3 MPa
  • Binding Capacity: >20mg DNA polymerase/ml
.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Quantity 1 mL
Regulatory Status RUO
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.