missing translation for 'onlineSavingsMsg'
Learn More

Heparin, MP Biomedicals™

Product Code. 11470672 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470672 250 mg 250mg
11440672 10 mg 10mg
11450672 50 mg 50mg
11460672 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11470672 Supplier MP Biomedicals Supplier No. 0219411483
  • ONLINE EXCLUSIVE PRICE
    274.00 €
    259.35 € / 250mg
    Save 14.65 €
    5% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Heparin is typically used as an anticoagulant, it binds to antithrombin III, a naturally occurring plasma protease inhibitor, accelerating the rate at which antithrombin III inhibits coagulation proteases (factor Xa and thrombin)

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689
Synonym Heparinic acid

Specifications

Color Colorless or White
Quantity 250 mg
Formula Weight ≈3000
Physical Form Powder or Solution
Chemical Name or Material Heparin, Sodium Salt from Porcine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.