missing translation for 'onlineSavingsMsg'
Learn More

Heparin, Sodium Salt from Porcine, MP Biomedicals™

Product Code. 11438282 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438282 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11438282 Supplier MP Biomedicals Supplier No. 0219411491
  • ONLINE EXCLUSIVE PRICE
    392.00 €
    371.70 € / 1g
    Save 20.30 €
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Heparin is a polymer classified as a mucopolysaccharide or a glycosoaminoglycan. It is biosynthesized and stored in mast cells of various mammalian tissues.

Product is made up of repeated disaccharide units consisting of 1,4-linked L-iduronic acid and D-glucosamine. The iduronic acid residues are O-sulfated at position 2, and the glucosamine residues are N-sulfated and O-sulfated at position 6. The repeated block can be interrupted or extended by residues of beta-D-glucuronic acid and 6-O-sulfated N-acetyl-alpha-D-glucosamine.

  • The amount of heparin needed to prevent coagulation in whole blood is between 20 units and 50 units per ml of whole blood

Specifications

CAS 9041-08-1
MDL Number MFCD00081689
Quantity 1 g
Synonym Heparinic acid
Formula Weight about 3,000g/mol
Physical Form Powder
compliance-icons
Product Identifier
  • Heparin Sodium Salt, Porcine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.