missing translation for 'onlineSavingsMsg'
Learn More

Heparin sodium

Product Code. 13478558 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 13478558

Brand: Thermo Scientific Acros 411210050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Heparin Sodium Salt, CAS # 9041-08-1, is derived from porcine intestinal mucosa and shows blood anti-coagulant activity. | CAS: 9041-08-1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Heparin sodium salt displays anti-coagulant activity. It can also efficiently inhibit mesenchymal cell proliferation and promotes the proliferation of primary epithelial cells
  • This compound can bind to antithrombin III and inhibit coagulation proteases

Applications

  • In in vitro studies, it is used as the main component of serum-free media to prevent cell aggregation
  • Heparin sodium salt can also induce the growth of human embryonic stem cells in a defined serum-free medium, acting as a cofactor for a basic fibroblast growth factor (FGF-2)
  • It is also applied in laboratory studies related to stem cell differentiation and growth
  • In in vitro studies this compound exhibits cytotoxic action inducing cancer cell death by interfering with transcription factor activity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689

Specifications

Absorbance ≤0.2% as nucleotides, 0.2 max. (0.4% in water) at 260nm, Protein and nucleotide impurities:, Protein and nucleotide impurities: at 0nm
Color White
pH Inclusive between 5.5 (1% soln.)
Ignition Residue Inclusive between 30%
Loss on Drying ≤8% (at 60°C for 4 hr.)
Infrared Spectrum Pass Test
Packaging Glass Bottle
Quantity 5 g
Merck Index 15, 4688
Percent Purity 150 I.U./mg min.
Concentration or Composition (by Analyte or Components) ≥150 I.U./mg
Physical Form Powder
Chemical Name or Material Heparin Sodium Salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.