missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanoic acid, 98%

Product Code. 11392286
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11392286

Brand: Thermo Scientific Alfa Aesar L05928.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 506-12-7 | C17H34O2 | 270.457 g/mol

Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.

Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly). Insoluble in water.

Notes
Store at 4°C. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-12-7
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00002751
InChI Key KEMQGTRYUADPNZ-UHFFFAOYSA-N
Synonym margaric acid, n-heptadecanoic acid, margarinic acid, n-heptadecylic acid, n-heptadecoic acid, heptadecylic acid, fatty acids, c16-18, unii-v987y9oz8l, normal-heptadecanoic acid, margarinate
PubChem CID 10465
ChEBI CHEBI:32365
IUPAC Name heptadecanoic acid
SMILES CCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 59°C to 63°C
Density 0.853
Boiling Point 227°C (100 mmHg)
Linear Formula CH3(CH2)15CO2H
Quantity 25 g
Beilstein 1781004
Merck Index 14,5751
Solubility Information Soluble in Chloroform (Slightly),Methanol (Slightly). Insoluble in water.
Formula Weight 270.46
Percent Purity 98%
Chemical Name or Material Heptadecanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Heptadecanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Heptadecanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.