missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanoic Acid, TRC

Product Code. 17999074
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17999074 5 g 5g
17969634 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17999074 Supplier Toronto Research Chemicals Supplier No. TRCH2810305G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 506-12-7
Molecular Formula C17 H34 O2
Molecular Weight (g/mol) 270.45
Synonym Heptadecanoic acid (8CI, 9CI, ACI), Margaric acid, Margarinic acid, n-Heptadecanoic acid, n-Heptadecoic acid, n-Heptadecylic acid, NSC 3743
IUPAC Name heptadecanoic acid
SMILES CCCCCCCCCCCCCCCCC(=O)O

Specifikationer

Quantity 5 g
InChI Formula InChI=1S/C17H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h2-16H2,1H3,(H,18,19)
Formula Weight 270.2559
Chemical Name or Material Margaric Acid
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.