missing translation for 'onlineSavingsMsg'
Learn More

Heptafluoro-1-iodopropane, 94%, stab. with copper

Product Code. 11452507
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452507 5 g 5g
11462507 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452507 Supplier Thermo Scientific Alfa Aesar Supplier No. A14784.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 754-34-7 | C3F7I | 295.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-34-7
Molecular Formula C3F7I
Molecular Weight (g/mol) 295.93
MDL Number MFCD00001061
InChI Key XTGYEAXBNRVNQU-UHFFFAOYSA-N
Synonym heptafluoro-1-iodopropane, heptafluoropropyl iodide, perfluoropropyl iodide, 1-iodoheptafluoropropane, 1-iodoperfluoropropane, propane, heptafluoro-1-iodo, 1-perfluoropropyl iodide, heptafluoroiodopropane, iodoheptafluoropropane, propane, heptafluoroiodo
PubChem CID 33977
IUPAC Name 1,1,1,2,2,3,3-heptafluoro-3-iodopropane
SMILES FC(F)(F)C(F)(F)C(F)(F)I

Specifications

Melting Point -95°C
Density 2.056
Boiling Point 40°C to 41°C
Refractive Index 1.329
Quantity 5 g
Beilstein 1758822
Sensitivity Light sensitive
Formula Weight 295.93
Percent Purity 97%
Chemical Name or Material Heptafluoro-1-iodopropane, stabilized with copper

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.