missing translation for 'onlineSavingsMsg'
Learn More

Heptafluoro-1-iodopropane, 94%, stab. with copper

Product Code. 11462507
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
missing translation for 'unitSize'
25g
5g
This item is not returnable. View return policy

Product Code. 11462507

missing translation for 'mfr': Thermo Scientific Alfa Aesar A14784.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 754-34-7 | C3F7I | 295.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-34-7
Molecular Formula C3F7I
Molecular Weight (g/mol) 295.93
MDL Number MFCD00001061
InChI Key XTGYEAXBNRVNQU-UHFFFAOYSA-N
Synonym heptafluoro-1-iodopropane, heptafluoropropyl iodide, perfluoropropyl iodide, 1-iodoheptafluoropropane, 1-iodoperfluoropropane, propane, heptafluoro-1-iodo, 1-perfluoropropyl iodide, heptafluoroiodopropane, iodoheptafluoropropane, propane, heptafluoroiodo
PubChem CID 33977
IUPAC Name 1,1,1,2,2,3,3-heptafluoro-3-iodopropane
SMILES FC(F)(F)C(F)(F)C(F)(F)I

Specifications

Melting Point -95°C
Density 2.056
Boiling Point 40°C to 41°C
Refractive Index 1.329
Quantity 25 g
Beilstein 1758822
Sensitivity Light sensitive
Formula Weight 295.93
Percent Purity 97%
Chemical Name or Material Heptafluoro-1-iodopropane, stabilized with copper

RUO – Research Use Only

Product Content Correction

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis