missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyramide, 97%

Product Code. 11406994
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406994 10 g 10g
11416994 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406994 Supplier Thermo Scientific Alfa Aesar Supplier No. B21466.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 662-50-0 | C4H2F7NO | 213.055 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 662-50-0
Molecular Formula C4H2F7NO
Molecular Weight (g/mol) 213.055
MDL Number MFCD00039773
InChI Key FOBJABJCODOMEO-UHFFFAOYSA-N
Synonym heptafluorobutyramide, perfluorobutyramide, butanamide, 2,2,3,3,4,4,4-heptafluoro, heptafluorobutanamide, heptafluorobutyroamide, heptafluoro-n-butyramide, butyramide, 2,2,3,3,4,4,4-heptafluoro, 2,2,3,3,4,4,4-heptafluorobutyramide, 2,2,3,3,4,4,4-heptafluoro-butyramide, butyramide, alpha,alpha,beta,beta,gamma,gamma,gamma-heptafluoro
PubChem CID 12626
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanamide
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)N

Specifications

Melting Point 102°C to 106°C
Quantity 10 g
Beilstein 1790527
Formula Weight 213.06
Percent Purity 97%
Chemical Name or Material Heptafluorobutyramide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.