missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric acid, 99%, for protein sequence analysis

Product Code. 13488558 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
150 mL
Packaging:
Glass Bottle
Unit Size:
100mL
150mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13488558 100 mL Glass Bottle 100mL
10674012 150 mL Glass Bottle 150mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13488558 Supplier Thermo Scientific Acros Supplier No. 411221000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 375-22-4 | C4HF7O2 | 214.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00004171
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

Absorbance (0.1% aqueous solution), (0.1% aqueous solution) at 0nm, 0.004 max. at 280nm, 0.006 max. at 254nm, 0.15 max. at 230nm
Melting Point -17.5°C
Color Beige to Brown-Orange
Density 1.6400g/mL
Boiling Point 120.0°C (755.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CF3CF2CF2CO2H
Refractive Index 1.3
Quantity 100 mL
UN Number 3265
Specific Gravity 1.64
Solubility Information Solubility in water: miscible. Other solubilities: soluble in benzene and carbon tetrachloride, miscible with acetone, ether and petroleum ether, insoluble in carbon disulfide and mineral oil
Formula Weight 214.04
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Heptafluorobutyric Acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.