missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric acid, 99%

Product Code. 11432717
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432717 100 g 100g
11422717 25 g 25g
11442717 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432717 Supplier Thermo Scientific Alfa Aesar Supplier No. A14907.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 375-22-4 | C4HF7O2 | 214.039 g/mol

Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.

Solubility
Miscible with water, toluene, ether, ethanol and benzene.       

Notes
Hygroscopic. Light sensitive. Incompatible withbases, oxidizing agents and reducing agents..   
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.039
MDL Number MFCD00004171
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

Melting Point -18°C
Density 1.648
Boiling Point 120°C to 121°C
Flash Point None
Odor Unpleasant
Linear Formula CF3CF2CF2CO2H
Refractive Index 1.3
Quantity 100 g
UN Number UN3265
Beilstein 1426882
Sensitivity Light sensitive
Solubility Information Miscible with water,toluene,ether,ethanol and benzene.
Formula Weight 214.04
Percent Purity 99%
Chemical Name or Material Heptafluorobutyric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.