missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric acid, 99%

Product Code. 10772021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772021 100 g Glass bottle 100g
10771831 25 g Glass bottle 25g
10639263 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10772021 Supplier Thermo Scientific Acros Supplier No. 172801000
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 375-22-4 | C4HF7O2 | 214.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4HF7O2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00004171
InChI Key YPJUNDFVDDCYIH-UHFFFAOYSA-N
Synonym heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
PubChem CID 9777
ChEBI CHEBI:39426
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

Melting Point -17.5°C
Color Colorless to Yellow
Density 1.6400g/mL
Boiling Point 120.0°C (755.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CF3CF2CF2CO2H
Refractive Index 1.3
Quantity 100 g
Specific Gravity 1.64
Solubility Information Solubility in water: miscible. Other solubilities: soluble in benzene and carbon tetrachloride, miscible with acetone, ether and petroleum ether, insoluble in carbon disulfide and mineral oil
Formula Weight 214.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Heptafluorobutyric acid
Show More Show Less
compliance-icons
Product Identifier
  • Heptafluorobutyric acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.