missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric Acid, TRC

Product Code. 30011046
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011046 5 g 5g
30012460 25 g 25g
17902585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30011046 Leverandør Toronto Research Chemicals Leverandørnr. TRCH2810405G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 375-22-4
Molecular Formula C4 H F7 O2
Molecular Weight (g/mol) 214.04
Synonym Butanoic acid, heptafluoro- (9CI), Butyric acid, heptafluoro- (6CI,8CI), 2,2,3,3,4,4,4-Heptafluorobutanoic acid, FC 23, Fluorad FC 23, H 0024, HFBA, Heptafluorobutanoic acid, Heptafluorobutyric acid, NSC 820, Perfluorobutanoic acid, Perfluorobutyric acid, Perfluoropropanecarboxylic acid, Butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, PFBA
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)F

Tekniske data

Quantity 5 g
InChI Formula InChI=1S/C4HF7O2/c5-2(6,1(12)13)3(7,8)4(9,10)11/h(H,12,13)
Formula Weight 213.9865
Percent Purity >95%
Chemical Name or Material Perfluorobutanoic acid
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.