missing translation for 'onlineSavingsMsg'
Learn More

Heptafluorobutyric Acid, TRC

Product Code. 30012460
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012460 25 g 25g
30011046 5 g 5g
17902585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012460 Supplier Toronto Research Chemicals Supplier No. TRCH28104025G
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-22-4
Molecular Formula C4 H F7 O2
Molecular Weight (g/mol) 214.04
Synonym Butanoic acid, heptafluoro- (9CI), Butyric acid, heptafluoro- (6CI,8CI), 2,2,3,3,4,4,4-Heptafluorobutanoic acid, FC 23, Fluorad FC 23, H 0024, HFBA, Heptafluorobutanoic acid, Heptafluorobutyric acid, NSC 820, Perfluorobutanoic acid, Perfluorobutyric acid, Perfluoropropanecarboxylic acid, Butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, PFBA
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4HF7O2/c5-2(6,1(12)13)3(7,8)4(9,10)11/h(H,12,13)
Formula Weight 213.9865
Percent Purity >95%
Chemical Name or Material Perfluorobutanoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.