missing translation for 'onlineSavingsMsg'
Learn More

Heptaldehyde, 95%, stabilized

Product Code. 10497110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10497110 50 mL Glass bottle 50mL
10214730 250 mL Glass bottle 250mL
10075903 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10497110 Supplier Thermo Scientific Acros Supplier No. 120320500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-71-7 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-71-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00007028
InChI Key FXHGMKSSBGDXIY-UHFFFAOYSA-N
Synonym heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
PubChem CID 8130
ChEBI CHEBI:34787
IUPAC Name heptanal
SMILES CCCCCCC=O

Specifications

Melting Point -42.0°C
Color Beige-Yellow to White
Density 0.8200g/mL
Boiling Point 150.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5CHO
Refractive Index 1.4115 to 1.4135
Quantity 50 mL
Beilstein 01, II, 750
Merck Index 15, 4693
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Stabilizer 100ppm hydroquinone
Viscosity 0.977 mPa.s (15°C)
Formula Weight 114.19
Percent Purity 95%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Heptaldehyde, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.