missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Heptane

Product Code. 15945600
Change view
Click to view available options
Quantity:
7 L
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15945600 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15945600 Supplier Riedel-de Haën Supplier No. 048827L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Heptane for preparative HPLC, ≥95.0% (GC).

  • Heptane
  • For preparative HPLC, ≥95.0% (GC)
  • n-Heptane, Heptanes
  • ≥95.0%, For preparative HPLC
  • CAS Number: 142-82-5
  • Linear Formula: CH3(CH2)5CH3
  • Molar Mass: 100.2 g/mol
  • Beilstein Registry Number: 1730763
  • EC Number: 205-563-8
  • MDL Number: MFCD00009544

Specifications

pH Not applicable
Melting Point -90.5°C
CAS 142-82-5
Color Colorless
CAS Max % ≥95.00%
Density 0.69 g/cm3 (15°C)
Quantity 7 L
Flash Point -4°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Molecular Formula C7H16
Linear Formula CH3(CH2)5CH3
Vapor Pressure 60.9 hPa (25°C)
MDL Number MFCD00009544
Viscosity No data available
UN Number UN1206
Beilstein 1730763
Concentration ≥95.00%
Synonym n-Heptane, Heptanes
Boiling Point 98.2°C to 98.4°C (1.000 hPa)
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
Percent Purity ≥95.00%
Grade HPLC
Chemical Name or Material Heptane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.