missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Heptane

Product Code. 15995610
Change view
Click to view available options
Quantity:
7 L
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15995610 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15995610 Supplier Riedel-de Haën Supplier No. 322877L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Heptane, Puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC).

  • All solvents are tested to meet ACS general use specifications.

Specifications

pH Not applicable
Melting Point -90.5°C
CAS 142-82-5
Color Colorless
CAS Max % 100.00%
Density 0.69 g/cm3 (15°C)
Quantity 7 L
Flash Point 6.7 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss. p.a.
Molecular Formula C7H16
Linear Formula CH3(CH2)5CH3
Vapor Pressure 60.9 hPa (25°C)
MDL Number MFCD00009544
Viscosity No data available
UN Number UN1206
Beilstein 1730763
Concentration ≥99.00%
Synonym n-Heptane, Heptanes
Boiling Point 98.2°C to 98.4°C (1.000 hPa)
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
Percent Purity ≥99.00%
Grade Pharmacopeia
Chemical Name or Material Heptane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.