Product Code. 15925490
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
15925490
7 L
7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
15925490
Supplier
Riedel-de Haën
Supplier No.
344957L
Please sign in to purchase this item. Need a web account? Register with us today!
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Heptane CHROMASOLV™ for pesticide residue analysis, ≥99%.
For gas chromatography (GC). Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC. Non-volatile matter and water content are specified for each solvent.
Specifications
pH
Not applicable
Melting Point
-90.5°C
CAS
142-82-5
Color
Colorless
Density
0.69 g/cm3 (15°C)
Boiling Point
98.2°C to 98.4°C (1.000 hPa)
Quantity
7 L
Flash Point
6.7 %(V)
Packaging
Returnable Stainless Steel Can
Physical Form
Liquid
Product Line
CHROMASOLV
Molecular Formula
C7H16
Linear Formula
CH3 (CH2 )5 CH3
MDL Number
MFCD00009544
Vapor Pressure
60.9 hPa (25°C)
UN Number
UN1206
Viscosity
No data available
Beilstein
1730763
Synonym
n-heptane, heptan
InChI Key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES
CCCCCCC
IUPAC Name
heptane
Molecular Weight (g/mol)
100.21
Percent Purity
≥99.00%
Grade
GC
Chemical Name or Material
Heptane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?