missing translation for 'onlineSavingsMsg'
Learn More

Heptane CHROMASOLV™, for HPLC, ≥99%, Solstice

Product Code. 15664650
Change view
Click to view available options
Quantity:
1 L
100 mL
2.5 L
2.5 L-PC
4 x 2.5 L
6 x 1 L
Pakningsstørrelse:
1 L
100mL
2.5 L
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
15664650 2.5 L 2.5 L
15634650 100 mL 100mL
15654650 1 L 1 L
15674650 2.5 L-PC 2.5 L
15694650 6 x 1 L Pack of 6
15684650 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Artikelnummer. 15664650 Leverandør Honeywell-Chromasolv Leverandørnr. 348732.5L
ChemsChrom Mega Deals
Use promo code 27993
Se alle kampagner
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Greener alternatives to Hexane in HPLC.

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Tekniske data

Melting Point -91°C
Density 0.684g/mL at 25°C
Quantity 2.5 L
Refractive Index n20/D 1.387
Linear Formula CH3(CH2)5CH3
Vapor Pressure 83mmHg (37.7°C)
Beilstein 1730763
Boiling Point 98°C
Formula Weight 100.2g/mol
Percent Purity ≥99%
Grade filtration through 0.2μm
Chemical Name or Material Heptane, for HPLC
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Heptane
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Informations supplémentaires
  • MIXTURE LIST-Contain: Heptane
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.