missing translation for 'onlineSavingsMsg'
Learn More

Heptane, Laboratory Reagent, Honeywell™

Product Code. 15647510 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 5 L
Packaging:
Fiberboard Box
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15647510 4 x 5 L Fiberboard Box Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15647510 Supplier Honeywell Research Chemicals Supplier No. 936414X5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Laboratory Reagent

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Especificaciones

CAS 142-82-5
Density 0.684g/mL at 25°C
Quantity 4 x 5 L
Explosion Limit 0.07
Flash Point −4°C (24.8°F)
Ignition Point 433°F
Packaging Fiberboard Box
Molecular Formula C7H16
Linear Formula CH3(CH2)5CH3
MDL Number MFCD00009544
Vapor Pressure 83mmHg (37.7°C), 40mmHg (20°C)
UN Number UN1206
Beilstein 1730763
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
PubChem CID 8900
ChEBI CHEBI:43098
Formula Weight 100.2g/mol
Chemical Name or Material Heptane
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.