missing translation for 'onlineSavingsMsg'
Learn More

Heptane (Peroxide-free/Sequencing), Fisher BioReagents

Product Code. 10479493
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10479493 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10479493 Supplier Fisher BioReagents Supplier No. BP1115500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Heptane is a solvent for other organics and is used as a carrier in chromatography.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -91°C
Absorbance 0.014 max. at 254nm, 0.2 max. (%) at 215nm, 0.75 max. at 200nm
CAS 142-82-5, 28729-52-4, 26635-64-3, 108-87-2
Color Colorless
Density 0.683g/cm³
Quantity 500 mL
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3845 to 1.3865 (at 25°C)
Residue after Evaporation 5ppm max.
Assay Percent Range ≥99 %
Molecular Formula C7H16
Linear Formula CH3(CH2)5CH3
Vapor Pressure 48mbar at 20°C
MDL Number MFCD00009544
Viscosity 0.4 mPa/s at 20°C
Water 0.02% max.
Merck Index 15, 4694
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
Boiling Point 98°C
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.21
PubChem CID 8900
ChEBI CHEBI:43098
Formula Weight 100.2
Percent Purity ≥99%
Grade Sequencing
Chemical Name or Material Heptane, Peroxide-free
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Heptane, anhydrous
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.